Digi-Sense® Temp10 Single-Input Thermocouple Thermometers, Cole-Parmer
Supplier: Antyila Scientific
91428-01 91428-00 91428-02
76292-160EA 927.14 CAD
76292-160 76292-162 76292-164
Digi-Sense® Temp10 Single-Input Thermocouple Thermometers, Cole-Parmer
Thermometers Digital Thermometers
Heavy-duty thermometer offers versatility and accuracy in a compact design.
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Low battery indicator
These robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealedMore Product Information