KIMBLE® ULTRA-WARE® Flat Bottom HPLC Reservoirs with Graduations, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953932-0252 953932-0502 953932-1002 953932-1003 953932-2002 953932-5002
KT953932-0252EA 210.67 CAD
KT953932-0252 KT953932-0502 KT953932-1002 KT953932-1003 KT953932-2002 KT953932-5002
KIMBLE® ULTRA-WARE® Flat Bottom HPLC Reservoirs with Graduations, DWK Life Sciences
Bottles HPLC Bottles
Designed for preparation, storage and delivery of all liquid chromatography mobile phases. As part of our ULTRA-WARE® series, these reservoirs are recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents.
  • Plastic safety-coating blocks UV light up to 385 nm, preventing photo-degradation of light-sensitive reagents
  • Safety coating helps to retain glass fragments and allows a reasonable amount of time for the safe disposal of liquid contents
  • Used with internal pressures up to 0.4 bar (6 psig) for helium sparging and
More Product Information