Kimble® Borosilicate Glass Tubes with ³/₄" Banded Labels, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60B12BZXW 60A10BZXB 60A10BZXG 60A10BZXL 60A10BZXW 60A10BZP 60B12BZP
89236-718CS 361.81 CAD
89236-718 89236-688 89236-690 89236-692 89236-694 10018-792 10018-794
Kimble® Borosilicate Glass Tubes with ³/₄" Banded Labels, DWK Life Sciences
Tubes Culture Tubes
These Kimble® glass culture tubes feature a permanent banded ceramic label, fused to the glass, which is easily written on with marker or pencil.
  • Permanent marking spots
  • Borosilicate glass tubes have excellent chemical resistance against sodium leaching
  • Labels are located ³/₄" from the open end and are available in a variety of colors
  • All tubes have sturdy, uniform bottoms and consistent lengths
Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.