Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination Units with Natural Refrigerants, Horizon Scientific
Supplier: Horizon Scientific
PRF092WWG/0 PRF092WWGLH/0 PRF092SSS/0 PRF092SSSLH/0 PRF072WWG/0 PRF072WWG/0A PRF122WWG/0 PRF122WWW/0 PRF122WWW/0A PRF122WWG/0A PRF092WWW/0 PRF092WWWLH/0
76300-576EA 6505.89 CAD
76300-576 76300-578 76300-580 76300-582 76300-584 76300-586 76300-636 76300-638 76300-640 76300-642 76300-644 76300-646
Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination Units with Natural Refrigerants, Horizon Scientific
Refrigerators
These efficient refrigerator and freezer combo units provide premier cooling for laboratory settings. They offer storage solutions for a variety of scientific and clinical applications.
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
Refrigerator units feature forced air directional cooling which provides superior temperature recovery after door openings. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.