Ammonium sulfate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Diammonium sulfate
, Sulphuric acid diammonium salt
, sulfuric acid diammonium salt
, Diammonium sulphate
, Mascagnite
Management of Change (MOC) category = RL
MACR3496-28
3496-28
CAMK3496-28EA
2898.79
CAD
MK3496-28
CAMK3496-28
Ammonium sulfate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™
Ammonium sulphate
Formula:
(NH₄)₂SO₄ MW: 132.14 g/mol Density: 1.77 g/cm³ |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 |
Specification Test Results
Meets F.C.C. Requirements | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
FCC - Assay ((NH₄)₂SO₄) | 99.0 - 100.5 % |
FCC - Arsenic (As) | ≤3 ppm |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Iron (Fe) | ≤0.0010 % |
FCC - Lead (Pb) | ≤3 mg/kg |
FCC - pH of Aqueous Solution | 4.5 - 6.0 |
FCC - Residue after Ignition | ≤0.25 % |
FCC - Selenium (Se) | ≤0.0030 % |
Solubility | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...