Sodium benzoate dense 99.01-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Benzoic acid sodium salt
Management of Change (MOC) category = R
MACR1331-26
MACR1331-26
MACR1331-26
CAMK1331-26EA
3690.66
CAD
MK1331-26
CAMK1331-26
JT1331-26
Sodium benzoate dense 99.01-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Sodium benzoate
Formula:
C₆H₅COONa MW: 144.11 g/mol Melting Pt: 410…430 °C Density: 1.44 g/cm³ Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012463 CAS Number: 532-32-1 EINECS: 208-534-8 Merck Index: 13,08654 |
Specification Test Results
Meets F.C.C. Requirements | |
Food GMP Manufactured Product | |
Meets N.F. Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
NF - Assay (C₆H₅COONa) (anhydrous basis) | 99.0 - 101.0 % |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Identification C | Passes Test |
NF - Alkalinity | Passes Test |
NF - Water (H₂O) | ≤1.5 % |
FCC - Assay (C₇H₅NaO₂) (anhydrous basis) | 99.0 - 101.0 % |
FCC - Alkalinity (as NaOH) | ≤0.04 % |
FCC - Arsenic (As) | ≤3 ppm |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Identification C | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Water (H₂O) | ≤1.5 % |
Solubility | Passes Test |
Appearance (white, crystalline granules) | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...