Michigan® Screw Machine Length Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
600LH-56-MD 600LH-43-MD 600LH-44-MD 600LH-32-MD 600LH-45-MD
76251-536EA 20.13 CAD
76251-536 76251-538 76251-540 76251-542 76251-544
Michigan® Screw Machine Length Drills, Cutting Tool Supply
Tool Sets
These drills are mainly for use in left hand spindle rotation on screw machines and drill heads.
  • 118° pointed
These can also be used as screw extractor for drilling out broken studs or fasteners. The shorter, rigid drills minimize deflection when heavy feed rates are used.