Bismuth(III) oxide ≥99.0% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Dibismuth trioxideBismuth Trioxide

Management of Change (MOC) category = N

J.T. Baker®
1159-R BAKR1159-R
CAJT1159-REA 17306.61 CAD
CAJT1159-R JT1159-R
Bismuth(III) oxide ≥99.0% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Bismuth (III) oxide
Formula: Bi₂O₃
MW: 465.96 g/mol
Melting Pt: 258…262 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 1304-76-3
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243

Order Now

Specification Test Results

For Microelectronic Use
Assay (as Bi₂O₃) (by EDTA titrn) ≥99.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.03 %
Sulfur Compounds (as SO₄) ≤0.002 %
Aluminum (Al) ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.005 %
Iron (Fe) ≤0.005 %
Lead (Pb) ≤0.005 %
Silicon (Si) ≤0.01 %
Sodium (Na) ≤0.01 %
Average Particle Diameter, µm (APD) ≤15
Particle Diameter - at 90%, For Information Only
Specific Surface Area, m²/g, For Information Only
Bulk Density (g/cc), For Information Only
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR