High Temperature Polyimide Labels for Thermal Transfer Printers, Brady
Supplier: Brady Worldwide
132469 139870
76249-270EA 3411.88 CAD
76249-270 76253-176
High Temperature Polyimide Labels for Thermal Transfer Printers, Brady
Labels
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Resists high temperatures up to 572 °F (300 °C)
  • Survives on the top or bottom side of printed circuit boards through the solder process
Withstands multiple cycles of harsh condition washes for printed circuit boards and electronic component preprocess labeling.
Certifications: Fulfills requirements for MIL-STD-202G Method 215K and SAE AS81531 Marking of Electrical InsulatingMore Product Information