J.T.Baker® DILUT-IT™, Sodium Phosphate, Dissolution Media Concentrate
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKRD021-00
BAKRD021-02
CAJTD021-0CS
1999.47
CAD
CAJTD021-0
CAJTD021-2
CAJTD021-4
J.T.Baker® DILUT-IT™, Sodium Phosphate, Dissolution Media Concentrate
Buffers
Biological Buffers
- pH at 25 °C at 26X Dilution: 6.75 to 6.85
- Store at room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...