Power Beams with Light Switch for Arlink® 8000 Workstations, Arlink
Supplier: ARLINK DIVISION CUST SERV TE
ARL-8647 ARL-8930 ARL-8860
76238-452EA 834.13 CAD
76238-452 76238-454 76268-298
Power Beams with Light Switch for Arlink® 8000 Workstations, Arlink
Furniture Tables and Workbenches
Provided with separate on/off switch for light, which allows remainder of beam to be left powered.
  • Single-sided configuration
  • 15 A standard
Mounts in micro slots, adjusts vertically in 1" increments.
Delivery information: Includes 7.5' power cord.