Bottoming Chamfer, Sid Tool
Supplier: SID TOOL CO INC TE
64832033 64047673
76251-130EA 191.61 CAD
76251-130 76251-132
Bottoming Chamfer, Sid Tool
Tool Sets
Bottoming chamfers are used for threading blind holes to the bottom. They have 1 to 2 threads. Bright provides a smooth, polished finish on the tool. It increases chip flow in softer materials such as aluminum, wood and plastic.
High Speed Steel (HSS) provides good wear-resistance and can be used in general purpose applications for both ferrous and non-ferrous materials.