Banana Test Connectors, Newark/Element
Supplier: NEWARK/ELEMENT 14 TE
108-0902-001 108-0903-001
76250-886EA 6.16 CAD
76250-886 76250-888
Banana Test Connectors, Newark/Element
Electrical Sockets
Banana jack with formed brass contact and nylon 6/6 body.
  • Bright tin finish
  • Nickel-plated nut hardware
  • 0.010Ω maximum contact resistance
  • Keyed mounting with double flats
Accepts up to 16 gauge wire.
Certifications: Nylon UL approved for self extinguishing.