J.T.Baker®, Cycle-Tap Samplers
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR9815-06 BAKR9815-01 BAKR9815-02 BAKR9815-03 BAKR9815-04 BAKR9815-05
CAJT9815-06CS 2656.86 CAD
CAJT9815-06 CAJT9815-1 CAJT9815-2 CAJT9815-3 CAJT9815-4 CAJT9815-5
J.T.Baker®, Cycle-Tap Samplers
Samplers Fluid Samplers
Cycle-Tap samplers are device for sampling small quantities of solvent.
These are designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.