Straight Flute Standard Hand Tap, Sid Tool
Supplier: SID TOOL CO INC TE
65156424 68581818
76251-910EA 59.27 CAD
76251-910 76251-912
Straight Flute Standard Hand Tap, Sid Tool
Tool Sets
High speed steel (HSS) provides good wear-resistance and can be used in general purpose applications for both ferrous and nonferrous materials.
Bottoming chamfers are used for threading blind holes. They have 1 to 2 threads.
Bright provides a smooth, polished finish on the tool. It increases chip flow in softer materials such as aluminum, wood and plastic.
Oxide, also known as black oxide or steam oxide, is a surface treatment that prevents chip building, galling and welding on the workpiece. It is commonly used on low carbons, stainless steel and ferrous metals.