Class 100 Accelerator Free Nitrile Gloves, TechNiGlove
Supplier: Techniglove
RVCR700 RVCR701 RVCR702 RVCR703 RVCR704 RVCR705
76302-802CS 502.81 CAD
76302-802 76302-804 76302-806 76302-808 76302-810 76302-812
Class 100 Accelerator Free Nitrile Gloves, TechNiGlove
Gloves
Rival CR 9.5" ambidextrous Class 100 nitrile gloves are soft modulus and accelerator-free for those with skin sensitivities.
  • Ambidextrous
  • Powder-free
Techniglove products are 100% nitrile, contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. Rival CR's new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort. Microtextured fingertips provide an excellent grip on all surfaces.
Suitable for use in cleanrooms and contamination controlled work environments, pharmaceutical, medical device, disc drive, and semiconductor manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag; 10 bags per case.