Engraving Cutters, Dremel
Supplier: DREMEL SERVICES INC. TE
DR-105 DR-106
76241-442EA 66.53 CAD
76241-442 76241-448
Engraving Cutters, Dremel
Cutters
Quality cutters for detail engraving, carving and routing in wood, fiberglass, plastic and soft metals.
  • Not for use with hardened materials
  • 1/8" Shank
  • Teal in color
These cutters need to use higher speeds for hard woods and slower speeds for metals.