Potassium nitrate 99.0-100.5%, granular USP, FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Nitrate of Potash
Management of Change (MOC) category = HR
3192-88
BAKR3192-88
CAJT3192-88CS
2334.59
CAD
JT3192-88
CAJT3192-88
Potassium nitrate 99.0-100.5%, granular USP, FCC, J.T.Baker®
Potassium nitrate
Formula:
KNO₃ MW: 101.1 g/mol Density: 2.10 g/cm³ |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 UN: 1486 ADR: 5.1,III |
Specification Test Results
Food GMP Manufactured Product | |
GMP Manufactured Product | |
Meets F.C.C. Requirements | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Assay (KNO₃) | 99.0 - 100.5 % |
USP - Arsenic (As) | ≤3 ppm |
USP - Chloride (Cl) | ≤0.03 % |
USP - Identification | Passes Test |
USP - Iron (Fe) | ≤0.001 % |
USP - Lead (Pb) (USP) | ≤0.001 % |
USP - Nitrite (NO₂) | ≤5 ppm |
USP - Sodium (Na) | ≤0.1 % |
USP - Sulfate (SO₄) | ≤0.1 % |
FCC - Assay (KNO₃) (dried basis) | 99.0 - 100.5 % |
FCC - Chlorate | Passes Test |
FCC - Identification | Passes Test |
FCC - Lead (Pb) (FCC) | ≤4 mg/kg |
FCC - Loss on Drying at 105°C | ≤1 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...