Glas-Col® Fabric Insulating Tops, Ace Glass
Supplier: Ace Glass
12047-210 12047-250 12047-270 12047-290 12047-26 12047-28
76291-794EA 827 CAD
76291-794 76291-796 76291-798 76291-800 76318-532 76318-534
Glas-Col® Fabric Insulating Tops, Ace Glass
Heaters Heating Mantles
Fabric insulating tops for 1, 2, and 3 neck flasks. Lacing closure allows installation without disturbing attached glassware or equipment. Insulating tops have no electrical connections.
Order Now
SPECIFICATIONS
Environmentally Preferable