CorRec-Pak® Shippers, Conductive Containers
Supplier: CONDUCTIVE CONTAINERS INC. TE
3500-10 3190-4 3500-10-2L
76248-324CS 51.21 CAD
76248-324 76248-352 76249-732
CorRec-Pak® Shippers, Conductive Containers
Mailers and Shippers
CorRec-Pak® Shippers are acceptable at static safe workstations, allowing the package to comply with your ESD. Regular Kraft corrugated can be insulative and generate damaging charges. Static dissipative convoluted urethane foam.
  • Top Foam Laminated, bottom foam loose
  • ESD safe circuit board shipping
  • Cost and labor saving compared to bags
  • Groundable at ESD workstation
These are ideal for ESD safe transport and storage of static sensitive circuit boards and assemblies.
Certifications: Compliance to ANSI/ESD S20.20 & ANIS/ESD-S541.