C470 Cartridge Tips for T470 and HT470 HD Models, JBC
Supplier: JBC TOOLS USA INC. TE
C470004 C470021 C470023
76248-538EA 386.45 CAD
76248-538 76253-108 76253-110
C470 Cartridge Tips for T470 and HT470 HD Models, JBC
Soldering Supplies Soldering Tips
C470 cartridges are the largest range featuring many shapes, sizes and special models.
  • Compatible with T470 HD Handles and HT470 HD Thermal Tweezers
Due to their size, these cartridges are appropriate for intensive soldering purposes and they are perfect for both SMD and applications with high power requirements.