Ammonium sulfate ≥99.0%, granular ACS, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

Management of Change (MOC) category = RL

BAKR3514-65 3514-F9
CAJT3514-65CS 34694.37 CAD
CAJT3514-65 JT3514-F9
Ammonium sulfate ≥99.0%, granular ACS, FCC, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Flash Pt: 1.77 g/cm³
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥99.0 %
ACS - Chloride (Cl) ≤3 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Phosphate (PO₄) ≤3 ppm
ACS - Residue on Ignition ≤0.005 %
Heavy Metals (as Pb) ≤3 ppm
Arsenic (As) ≤0.200 ppm
Solution Test Passes Test
FCC - Assay ((NH₄)₂SO₄)(F.C.C.) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤3 mg/kg
FCC - Residue on Ignition (FCC) ≤0.25 %
FCC - Selenium (Se) ≤0.003 %
Loss on Drying at 105°C ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR