Loose Standard Tissue Processing/Embedding Cassettes with Lid, Avantik
Supplier: Avantik
EM0169-10 EM0169-11 EM0169-12 EM0169-14 EM0169-2 EM0169-3 EM0169-4 EM0169-5 EM0169-6 EM0169-7 EM0169-8 EM0169-9
76269-684CS 637.14 CAD
76269-684 76269-686 76270-414 76270-416 76270-418 76270-420 76270-422 76270-424 76270-426 76270-428 76270-430 76270-432
Loose Standard Tissue Processing/Embedding Cassettes with Lid, Avantik
Embedding Cassettes Tissue Processing Cassettes
Avantik boxed loose standard tissue processing/embedding cassettes with lid are made with the highest quality and provide excellent value.
  • Anterior writing area is at a 35° angle
  • Three-sided writing surface
Molded from a high density polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap latch and hinge-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable