Stackable Metal Slide Cabinets, Avantik
Supplier: Avantik
ST7076 ST7077
76270-350EA 779.49 CAD
76270-350 76270-352
Stackable Metal Slide Cabinets, Avantik
Microscope Slide Boxes
Holds up to 5000 standard 3×1" microscope slides.
  • Interlocking, stackable to 10 cabinets high
  • Cabinet and base are stackable with the Boekel Cabinet
  • Cabinet base features rubber feet to prevent scratching and slipping
Cabinet is supplied with a two-year warranty.
Delivery information: Includes plastic drawer dividers, sponge blocks spaces and identification labels.