D-(+)-Sucrose, GenAR® NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

Management of Change (MOC) category = R

MACR7724-88 7724-88
CAMK772488EA 1449.02 CAD
CAMK772488 MK772488
D-(+)-Sucrose, GenAR® NF, Macron Fine Chemicals™
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://ca.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Store in a tightly closed container.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Appearance of Solution Passes Test
NF - Color Value ≤45
NF - Conductivity ≤35 µS/cm
NF - Optical Rotation (+) 66.3 - 67.0 Degree
NF - Reducing Sugars Passes Test
NF - Sulfite (SO2) ≤10 ppm
NF - Loss on Drying at 105 ºC ≤0.1 %
EP/BP - Identification A Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Color Value ≤45
EP/BP - Conductivity, µS cm⁻¹ ≤35
EP/BP - Optical Rotation (+) 66.3 - 67.0 Degree
EP/BP - Sulfite (SO₂) ≤10 ppm
EP/BP - Loss on Drying at 105°C ≤0.1 %
Endotoxin Concentration (EU/g) ≤250
EP/BP - Reducing Sugars Passes Test
Additional Tests - Dextrins Passes Test
Additional Tests - Lead (Pb) ≤0.5 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR