Sodium carbonate monohydrate ≥99.5%, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Sodium carbonate hydrous, Soda monohydrate

Management of Change (MOC) category = N

BAKR3598-07 BAKR3598-09
CAJT3598-7EA 2777.29 CAD
CAJT3598-7 CAJT3598-9 CAJT3598-69
Sodium carbonate monohydrate ≥99.5%, BAKER ANALYZED® ACS
Sodium carbonate monohydrate
Formula: Na₂CO₃·1H₂O
Density: 2.25 g/cm³
MDL Number: MFCD00149177
CAS Number: 5968-11-6

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂CO₃·H₂O) ≥99.5 %
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Drying at 150°C 13.0 - 15.0 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Calcium (Ca) ≤0.03 %
ACS - Silica (SiO₂) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.004 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.005 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR