Ammonium sulfate 99.0-100.5%, granular NF, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Diammonium sulfateSulphuric acid diammonium saltsulfuric acid diammonium saltDiammonium sulphateMascagnite

Management of Change (MOC) category = R

J.T. Baker®
3523-65 BAKR3523-65
CAJT3523-65EA 15710.52 CAD
CAJT3523-65 JT3523-65
Ammonium sulfate 99.0-100.5%, granular NF, ACS, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Density: 1.77 g/cm³
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Specification Test Results

Meets N.F. Requirements
GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
NF - Insoluble Matter ≤0.005 %
NF - Residue on Ignition ≤0.005 %
NF - pH of 5% Solution 5.0 - 6.0
NF - Nitrate (NO₃) ≤0.001 %
NF - Chloride (Cl) ≤5 ppm
NF - Phosphate (PO₄) ≤5 ppm
NF - Iron (Fe) ≤5 ppm
NF - Identification Passes Test
NF - Total Aerobic Microbial Count ≤100 cfu/g
NF - Total Yeast and Mold Count ≤10 cfu/g
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥99.0 %
ACS - Residue on Ignition ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤3 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - Phosphate (PO₄) ≤3 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
Appearance (White crystals or granules) Passes Test
No Class 1, 2 or 3 Residual Solvents are likely to be present by
exclusion of the production/purification process.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Preserve in well closed container.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR