J.T.Baker® BAKERBOND®, PolyPEI Multimode Ion Exchange Chromatography Resin
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR7585-03 BAKR7585-04 BAKR7585-05
CAJT7585-03CS 16162.86 CAD
CAJT7585-03 CAJT7585-04 CAJT7585-05
J.T.Baker® BAKERBOND®, PolyPEI Multimode Ion Exchange Chromatography Resin
Chromatography Sorbents HPLC Sorbents
The J.T.Baker® BAKERBOND® PolyPEI multimode ion exchange (IEX) chromatography resin functions as a weak anion exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining weak anion exchange functionality with hydrophilicity, BAKERBOND® PolyPEI multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyPEI ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Ordering information: 1 mL and 5 mL lab columns are also available under complementary products.