Black Oxide Pin Gages Set, Zoro
Supplier: ZORO - CUSTOMER SERVICE TE
901200300 901200600 901200500
76249-394EA 1511.13 CAD
76249-394 76249-542 76249-720
Black Oxide Pin Gages Set, Zoro
Pin Gage Set, Minus, Cylindrical, Class ZZ.
  • Black oxide coated
  • Made of 52100 tool steel - 60/62 Rc
The gage has accuracy of +0.0000/–0.0002" and increaments of 0.001".
Delivery information: Includes Certificate of Accuracy with NIST traceability.