Clamp Gaskets, Flanged, Garlock Hygienic Technologies
Supplier: Rubber Fab
40MPFSFY-300 40MPFSFY-400
76239-920EA 24.47 CAD
76239-920 76239-922
Clamp Gaskets, Flanged, Garlock Hygienic Technologies
Seals Gaskets
Sanitary Tri-Clamp® Type II Flanged Gaskets are used widely in the Food, Dairy, Beverage, Biotech, Pharmaceutical, and all other Sanitary Process industries to seal clamp connections in sanitary pipe lines.
  • Seals pressures to flange ratings, in accordance with ASME B16.5 and B16.47
  • Inner ring can help shield sealing elements from erosion caused by abrasive media
  • Peroxide cured
  • Not recommended for use with bolted clamps
These Type II Flanged Sanitary Gaskets are manufactured with a flanged design to aid in installation.
Certifications: Meets 21CFR 177.2600; passed USP Class VI Cytotoxicity testing.