jetOPTIMUS® DNA transfection Reagent
Supplier: Polyplus Transfection
jetOPTIMUS® is a best-in-class DNA transfection reagent, based on a novel cationic nanotechnology allowing high efficiency with low volume of reagent. Reach higher transfection efficiency while doubling the number of transfection per vial!
- Highly efficient: reach maximal gene expression in hard-to-transfect cells
- Cost-effective: use minimum reagent volume and DNA quantity
- Biologically relevant: keep an excellent cell viability & morphology
- Time-saving: transfect with an optimized ready-to-use protocol
jetOPTIMUS® transfection reagent improves cellular uptake and endosomal escape of DNA in cells (even in hard-to-transfect cells) resulting in higher transfection efficiency. In order to work in relevant physiological conditions, transfection with jetOPTIMUS® requires a minimum DNA quantity and reagent volume to keep an excellent cell viability and morphology.
Certifications: Free of charge samples are available upon request. Please contact your local Avantor sales office for further details. Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.