Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

BAKR7451-07 BAKR7451-26
CAJT7451-07CS 780.3 CAD
CAJT7451-07 CAJT7451-26
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®
Sodium hydrogen carbonate

Caution: This product is not intended for use in hemodialysis.

Formula: NaHCO₃
Density: 2.20 g/cm³
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
ACS - Ammonium (NH₄) ≤5 ppm
ACS - Assay (NaHCO₃) (by acidimetry) 99.7 - 100.3 %
ACS - Calcium (Ca) ≤0.02 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.015 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Chloride (Cl) ≤0.015 %
USP - Identification Passes Test
USP - Insoluble Substances Passes Test
USP - Limit of Ammonia ≤20 ppm
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Normal Carbonate Passes Test
USP - Arsenic (As) ≤2 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
This product is not intended for use in hemodialysis.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR