Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = R
BAKR5683-07
BAKR5683-08
BAKR5683-09
CAJT5683-7EA
2174.96
CAD
CAJT5683-7
CAJT5683-08
CAJT5683-09
Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®
Orthophosphoric acid
Formula:
H₃PO₄ Density: 1.05 g/cm³ |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 UN: 1805 ADR: 8,III |
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Alkali Phosphates | Passes Test |
NF - Identification | Passes Test |
NF - Nitrate (NO₃) | Passes Test |
NF - Phosphorous or Hypophosphorous Acid | Passes Test |
NF - Sulfate (SO₄) | Passes Test |
NF - Assay (as H₃PO₄), g/100 mL | 9.5 - 10.5 |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Chloride (Cl) | ≤ 6 ppm |
EP/BP - Phosphorous or Hypophosphorous Acid | Passes Test |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Iron (Fe) | ≤ 6 ppm |
EP/BP - Substances Precipitated with Ammonia | Passes Test |
EP/BP - Sulfate (SO₄) | ≤ 10 ppm |
EP/BP - Assay (H₃PO₄) | 9.5 - 10.5 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...