1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemical
MB-081-0100 MB-081-1000
76236-542EA 134.3 CAD
76236-542 76236-544
1X PBS Buffer with 1% Casein
Buffers Electrophoresis Buffers Washing Buffers
Phosphate buffered saline with casein is suitable for multiple applications including biological diluent buffer for antibodies or other biologics.
  • Ready-to-Use
  • Diluent or wash buffer for assays
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
It also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
Application Note: This product is a ready-to-use 1X solution and no further dilution is required.