10X PBS pH 7.2

Supplier: Rockland Immunochemicals
MB-008-4000
76236-390EA 293.88 CAD
76236-390
10X PBS pH 7.2
Buffers Electrophoresis Buffers Washing Buffers
10X Phosphate buffered saline (PBS) pH 7.2

  • Concentrated 10X
  • 0.2 M Potassium Phosphate, 1.5 M Sodium Chloride, pH 7.2
  • Buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, ELISA, and more.

Phosphate buffered saline is suitable for multiple applications including biological diluent buffer for antibodies, other biologics, and washes. Other applications may require detergents or other additional components.

Application Note: This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. 10X Phosphate Buffered Saline (PBS) consists of 0.2 M Potassium Phosphate, 1.5 M Sodium Chloride, pH 7.2 prepared in highly polished pharmaceutical grade water (WFI).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR