J.T.Baker® BAKERBOND®, PolyABx Multimode Ion Exchange Chromatography Resin
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
The J.T.Baker® BAKERBOND® PolyABx multimode ion exchange (IEX) chromatography resin functions as a weak cation exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining weak cation exchange functionality with weak anionic exchange sites and hydrophilicity, BAKERBOND® PolyABx multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyABx ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.