Kimble® Desiccators with Plastic Top Knob, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
31050-150 31050-250 31050-200
75792-272CS 831.27 CAD
75792-272 75792-200 75792-274
Kimble® Desiccators with Plastic Top Knob, DWK Life Sciences
Desiccators Desiccators, Glass
Excellent for storage of hygroscopic chemicals, and to prevent adsorption of moisture when using crucibles, weighing bottles, and watch glasses.
  • Plastic screw-thread top cap for easy cover removal
  • Recessed base for containment of desiccant
  • Includes metal plate
  • Glass bottom and lid
Certifications: Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II, requirements.