6 Position TrueBlot® Magnetic Separator, Rockland Immunochemicals
Supplier: Rockland Immunochemical
TMS-06 TMS-15-50 TMS-32
76237-652EA 861.1 CAD
76237-652 76237-654 76237-656
6 Position TrueBlot® Magnetic Separator, Rockland Immunochemicals
Beads Magnetic Beads
6x1.5mL Position Magnetic Separator
  • magnetic separation
  • six 1.5 mL microcentrifuge tubes
A magnetic separation unit designed to accommodate up to six 1.5 mL microcentrifuge tubes in order to separate magnetic particles contained in those tubes. Optimal working volume is up to 1 mL.
Magnetic separator unit can be used to hold, and separate magnetic particles.
Order Now
SPECIFICATIONS
Environmentally Preferable
Shipping conditions At ambient temperatures