HRP Conjugate Stabilizer, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-076-1000 MB-076-0100
76236-530EA 154.84 CAD
76236-530 76236-532
HRP Conjugate Stabilizer, Rockland Immunochemicals
Assays Immunological Assays
HRP Conjugate Stabilizer
  • Ready-to-Use
  • azide and mercury free
  • use with Horseradish Peroxidase (HRP) conjugated primary or secondary antibodies
  • suitable in assays or as buffer, not recommended for use with luminol based assays
HRP Conjugate Stabilizer is to be used with Horseradish Peroxidase (HRP) conjugated primary or secondary antibodies or other proteins or compounds in solution.   The components of the stabilizer (proprietary) are unique, and stabilize both the antibody and enzyme portions of the conjugate. HRP Conjugate Stabilizer prevents the loss of catalytic activity and maintains the active conformation of the antibody portion of the conjugate.
This product is a ''ready-to-use'' 1X diluent for Horseradish Peroxidase (HRP) conjugated primary or secondary. This buffer contains buffered saline, and proprietary reagents to stabilize and preserve Horseradish Peroxidase (HRP) conjugated primary or secondary antibodies in solution. A proprietary combination of preservatives is used that is azide and mercury free. It is suitable for ELISA, Western Blot, Southern/Northern Blots, and lateral flow or directly as an assay buffer. This buffer is not recommended to be used in Luminol based assays.