Mouse TIMP-4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0802
76236-192EA 1150.24 CAD
76236-192
Mouse TIMP-4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TIMP-4 AccuSignal ELISA Kit
  • Natural and recombinant mouse TIMP-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: C30-P224
Metalloproteinase inhibitor 4 is an enzyme that in humans is encoded by the TIMP4 gene. This gene belongs to the tissue inhibitor of metalloproteinases gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. TIMP4 gene is mapped to chromosome 3p25. TIMP4 gene contains 5 exons that span 6 kb of genomic DNA. The gene is expressed as a 1.4-kb transcript abundant in heart and present at low levels in several other tissues. Expressed TIMP4 inhibits MMPs in vitro. What’s more, TIMP-4 binds both progelatinase A and the C domain in a similar manner to that of TIMP-2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5,000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml mouse TIMP-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse TIMP-4 standard solution and each sample is measured in duplicate.