Human Cathepsin E ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0780
76236-172EA 1150.24 CAD
76236-172
Human Cathepsin E ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Cathepsin E AccuSignal ELISA Kit
  • Natural and recombinant human Cathepsin E.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q18-P396
Cathepsin E is a protein that in humans is encoded by the CTSE gene. It is mapped to 1q32.1. The protein encoded by this gene is a gastric aspartyl protease that functions as a disulfide-linked homodimer. This protease, which is a member of the peptidase C1 family, has a specificity similar to that of pepsin A and cathepsin D. It is an intracellular proteinase that does not appear to be involved in the digestion of dietary protein and is found in highest concentration in the surface of epithelial mucus-producing cells of the stomach. Cathepsin E is the first aspartic proteinase expressed in the fetal stomach and is found in more than half of gastric cancers. It appears, therefore, to be an oncofetal antigen, and it also has an important role in immune responses.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Cathepsin E standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Cathepsin E standard solution and each sample be measured in duplicate.