Mouse CD200 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0684
76235-848EA 976.44 CAD
76235-848
Mouse CD200 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD200 AccuSignal ELISA Kit
  • Natural and recombinant mouse CD200.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q31-G232
OX-2 membrane glycoprotein, also known as CD200 is a human protein encoded by the CD200 gene. This gene is mapped to 3q13.2. The protein encoded by this gene is a type-1 membrane glycoprotein, which contains two immunoglobulin domains, and thus belongs to the immunoglobulin superfamily. Studies of the related genes in mouse and rat suggest that this gene may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. It is suggested that CD200-CD200R signaling plays a central role in macrophage fusion and osteoclast formation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse CD200 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD200 standard solution and each sample be measured in duplicate.