Mouse IL1RL1 - ST2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0680
76235-840EA 1055.44 CAD
76235-840
Mouse IL1RL1 - ST2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse IL1RL1 - ST2 AccuSignal ELISA Kit
  • Natural and recombinant mouse IL1RL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S27-R332
Interleukin 1 receptor-like 1, also known as IL1RL1 or ST2, is a protein that in humans is encoded by the IL1RL1 gene. The protein encoded by this gene is a member of the interleukin 1 receptor family. Studies of the similar gene in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. TL1RL1 is necessary for endotoxin tolerance and, by inhibiting TLR responses, enhances Th2 responses. This gene, interleukin 1 receptor, type I(IL1R1), interleukin 1 receptor, type II(IL1R2) and interleukin 1 receptor-like 2(IL1RL2) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse IL1RL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL1RL1 standard solution and each sample be measured in duplicate.