Mouse TNFRSF13B - TACI ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0656
76236-942EA 1150.24 CAD
76236-942
Mouse TNFRSF13B - TACI ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TNFRSF13B - TACI AccuSignal ELISA Kit
  • Natural and recombinant mouse TNFRSF13B.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A2-C128
Tumor necrosis factor receptor superfamily member 13B, also known as TNFRSF13B or more commonly as TACI, is a transmembrane receptor protein found predominantly on the surface of B cells, which are an important part of the immune system. TACI controls T cell-independent B cell antibody responses, isotype switching, and B cell homeostasis. TACI is a lymphocyte-specific member of the tumor necrosis factor(TNF) receptor superfamily. It was originally discovered because of its ability to interact with calcium-modulator and cyclophilin ligand(CAML). TACI was later found to play a crucial role in humoral immunity by interacting with two members of the TNF family: BAFF and APRIL. These proteins signal through TACI inducing activation of several transcription factors including NFAT, AP-1, and NF-kappa-B which then modulate cellular activities. Defects in the function of TACI can lead to immune system diseases.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 188pg/ml, 94pg/ml, 46.9pg/ml mouse TNFRSF13B standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse TNFRSF13B standard solution and each sample is measured in duplicate.