Human Fetuin A ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0448
76235-614EA 1150.24 CAD
76235-614
Human Fetuin A ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Fetuin A AccuSignal ELISA Kit
  • Natural and recombinant human Fetuin A.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A19-V367
Alpha2-HS glycoprotein(AHSG), also known as fetuin-A, is a plasma protein displaying high-affinity interaction with calcium phosphate, by which ectopic vascular calcification is prevented. The AHSG polymorphism is attributable to the hereditary variation of AHSG and phosphate serum levels, which may affect skeletal development and chronic disorders such as vascular calcification.1Human plasma protein alpha 2-HS-glycoprotein(AHSG) is composed of two polypeptide chains, A and B, encoded by a single mRNA. Southern blot analysis of mouse x human somatic cell hybrids has mapped the AHSG gene to human chromosome 3 in the region 3q21----qter. Using a recombinant plasmid containing a 1,538 bp insert spanning the entire AHSG coding region, AHSG was localized to chromosomal bands 3q27----q29 by in situ hybridization.2
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human Fetuin A standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA) to each empty well. It is recommended that each human Fetuin A standard solution and each sample be measured in duplicate.