Human CXCL7 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0426
76235-586EA 1704.82 CAD
76235-586
Human CXCL7 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CXCL7 AccuSignal ELISA Kit
  • Natural and recombinant human CXCL7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A59-D128
Chemokine(C-X-C motif) ligand 7(CXCL7), also called PPBP or SCYB7, is a human gene. The encoded protein, Chemokine(C-X-C motif) ligand is a small cytokine belonging to the CXC chemokine family. CXCL7 is mapped to 4q13.3. It is a protein that is released in large amounts from platelets following their activation. CXCL7 is the precursor of the 2 platelet alpha-granule proteins, platelet basic protein(PBP) and connective tissue-activating peptide III(CTAP3). It stimulates various processes including mitogenesis, synthesis of extracellular matrix, glucose metabolism and synthesis of plasminogen activator. It also stimulates the formation and secretion of plasminogen activator by synovial cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CXCL7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each human CXCL7 standard solution and each sample be measured in duplicate.