Mouse B7-1 - CD80 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0411
76236-888EA 1150.24 CAD
76236-888
Mouse B7-1 - CD80 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse B7-1 - CD80 AccuSignal ELISA Kit
  • Natural and recombinant human CD80.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D37-K245
The protein CD80(Cluster of Differentiation 80) is a molecule found on activated B cells and monocytes which provides a costimulatory signal necessary for T cell activation and survival. It is also known as B7.1. The cDNA for B7-1 predicts a type I membrane protein, i.e., one synthesized with a signal peptide that is cleaved upon translocation across the endoplasmic membrane. The protein is predicted to contain 2 extracellular domains structurally similar to those of Ig, a hydrophobic transmembrane region, and a short cytoplasmic domain. The CD80 and CD86(601020) genes encode B7-1 and B7-2, respectively, which are structurally similar members of the immunoglobulin superfamily expressed on a variety of hematopoietic cell types. stated that B7-1 and B7-2 provide a costimulatory signal to T cells by interacting with CD28 and CTLA4. The standard product used in this kit is recombinant B7-1, D37—K245, which is composed of two single chains acids with the dipolymer.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse CD80 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse CD80 standard solution and each sample be measured in duplicate.