Human CD36 - SR-B3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0405
76237-268EA 1150.24 CAD
76237-268
Human CD36 - SR-B3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD36 - SR-B3 AccuSignal ELISA Kit
  • Natural and recombinant human CD36.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: G30-N439
CD36(Cluster of Differentiation 36) is an integral membrane protein found on the surface of many cell types in vertebrate animals and is also known as FAT, SCARB3, GP88, glycoprotein IV(gpIV) and glycoprotein IIIb(gpIIIb). The human CD36 is a member of a gene family of structurally related glycoproteins and functions as a receptor for collagen type I and thrombospondin. The use of a CD36 genomic probe has allowed the localization of the CD36 locus to the 7q11.2 band by fluorescence in situ hybridization coupled with GTG-banding. CED-1/SCARF1 and C03F11.3/CD36 are beta-glucan binding receptors and define an evolutionarily conserved pathway for the innate sensing of fungal pathogens.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CD36 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD36 standard solution and each sample be measured in duplicate.