Human EGFR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0158
76237-128EA 1150.24 CAD
76237-128
Human EGFR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human EGFR AccuSignal ELISA Kit
  • Natural and recombinant human EGFR.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-S645
The epidermal growth factor receptor(EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family(EGF-family) of extracellular protein ligands.1 It is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR(ErbB-1), HER2/c-neu(ErbB-2), Her 3(ErbB-3) and Her 4(ErbB-4). EGFR exists on the cell surface and is activated by binding of its specific ligands, including epidermal growth factor and transforming growth factor alpha(TGFalpha). EGFR and its ligands are cell signaling molecules involved in diverse cellular functions, including cell proliferation, differentiation, motility, and survival, and in tissue development. Mutations that lead to EGFR overexpression(known as upregulation) or overactivity have been associated with a number of cancers, including lung cancer and glioblastoma multiforme. In this latter case a more or less specific mutation of EGFR, called EGFRvIII is often observed.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml human EGFR standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA) or human milk to each empty well. It is recommended that each human EGFR standard solution and each sample be measured in duplicate.