RAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D110-10-0100 D110-10-0500 D110-10-1000
76226-480EA 1594.22 CAD
76226-480 76226-482 76226-484
RAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Sera (Serum)
Heat Inactivated Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Rat Serum is used as a supplement to cell culture media. Rat Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Rat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Rat Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal